Learn More
Grainger pH Insertion Assembly CPVC BNC Flat Tip
Catalog No. 502768096
Encompass_Preferred
The Grainger 16Z021 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for light duty scrubbing and cleaning. It measures 6 inches by 9 inches and features an open web design that rinses easily. The pad is made of high-quality fibers that are non-scratching on most surfaces. It can be used wet or dry on a variety of materials. The 3M Scotch-Brite pad is ideal for general purpose cleaning in industrial, food service, and commercial settings. It provides an economical and effective cleaning solution.
Catalog No. 50-276-8096 Supplier Grainger Supplier No. 16Z021
The Grainger 16Z021 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for light duty scrubbing and cleaning. It measures 6 inches by 9 inches and features an open web design that rinses easily. The pad is made of high-quality fibers that are non-scratching on most surfaces. It can be used wet or dry on a variety of materials. The 3M Scotch-Brite pad is ideal for general purpose cleaning in industrial, food service, and commercial settings. It provides an economical and effective cleaning solution.