Learn More
Hach Company Intellical™ PHC301 Laboratory General Purposes Refillable pH Electrode, 3 m Cable
Catalog No. 501998081 Shop All Hach Company Products
Encompass
Intellical™ PHC301 is a digital combination pH electrode with built-in temperature sensor. The PHC301 has a refillable with two ceramic pins reference junction. This laboratory pH electrode is shockproff with its Zeonor plastic body protecting down to the glass bulb sensing element. The electrode is available with a 3 or 1 m cable. The PHC301 refillable combination pH electrode is recommended for wastewater, drinking water and general aquous applications. The PHC301 is not suitable for use with organic solvents. A 28-mL bottle of reference electrolyte filling solution (3 M KCl solution saturated with AgCl) is included with the probe.
Catalog No. 50-199-8081 Supplier Hach Company Supplier No. PHC30103
Intellical™ PHC301 is a digital combination pH electrode with built-in temperature sensor. The PHC301 has a refillable with two ceramic pins reference junction. This laboratory pH electrode is shockproff with its Zeonor plastic body protecting down to the glass bulb sensing element. The electrode is available with a 3 or 1 m cable. The PHC301 refillable combination pH electrode is recommended for wastewater, drinking water and general aquous applications. The PHC301 is not suitable for use with organic solvents. A 28-mL bottle of reference electrolyte filling solution (3 M KCl solution saturated with AgCl) is included with the probe.