Learn More
Cole-Parmer Jenway 3520 Advanced Digital pH Meter Kit w/GLP, glass electrode, ATC, buffers; 120 V
Catalog No. 501961253 Shop All Cole-Parmer Products
Encompass_Preferred
Benchtop pH meters feature up to three decimal place resolution, support up to three pH calibration points, and offer manual and automatic buffer recognition for DIN, JIS, and NIST buffers. A large LCD displays status and results of pH and temperature readings. Data is easily exported to a computer or printer via RS-232 connection.
Catalog No. 50-196-1253 Supplier Cole-Parmer Supplier No. 3545910
Benchtop pH meters feature up to three decimal place resolution, support up to three pH calibration points, and offer manual and automatic buffer recognition for DIN, JIS, and NIST buffers. A large LCD displays status and results of pH and temperature readings. Data is easily exported to a computer or printer via RS-232 connection.