Learn More
PCE AMERICAS INC Water Analyser with GLP data management -2 … +20 pH.
SDP
Catalog No. NC3654416
Encompass_Preferred
The environmental meter PCE-228 is a very easy-to-use pH / mV / °C / °F handheld instrument. The pH value and the temperature can be stored directly on the SD card (Excel file) or transferred directly from the environmental meter to a PC via the RS-232 interface. For this we optionally offer a software and the suitable RS-232 interface cable as an accessory. Temperature compensation is either manual or automatic via the supplied temperature sensor.
Catalog No. NC3654416 Supplier PCE AMERICAS INC Supplier No. PCE228LIQ
The environmental meter PCE-228 is a very easy-to-use pH / mV / °C / °F handheld instrument. The pH value and the temperature can be stored directly on the SD card (Excel file) or transferred directly from the environmental meter to a PC via the RS-232 interface. For this we optionally offer a software and the suitable RS-232 interface cable as an accessory. Temperature compensation is either manual or automatic via the supplied temperature sensor.