Learn More
PCE AMERICAS INC Portable pH meter that measure pH, ORP & pH. Temperature
SDP
Supplier:  PCE AMERICAS INC PCE228SLUR
Encompass_Preferred
"The water analysis meter PCE-228 is a very easy-to-use pH / mV / °C / °F handheld instrument. The pH value and the temperature can be stored directly on the SD card (Excel file) or transferred via the RS-232 interface directly from the water analysis meter to a PC. For this we optionally offer a software and the suitable RS-232 interface cable as an accessory. Temperature compensation is either manual or automatic via the supplied temperature sensor.All this allows a fast and reliable pH measurement. The water analysis meter has a three-point calibration, which is performed automatically. It can be determined with the combination meter, the pH, the temperature or the REDOX potential (ORP). For the latter measurement parameters, however, a REDOX electrode must also be ordered. Here we recommend the ORP electrode ORP-14. The water analysis meter is battery powered (6 x 1.5V batteries are included)."
Catalog No. NC3654424