Learn More
Electron Microscopy Sciences Basic pH Meter Kit
SDP
Catalog No. 501929321
Encompass_Preferred
Includes everything necessary for immediate field use: Basic pH Meter, epoxy body, sealed reference combination electrode with a BNC connector and 2.5-ft cable, pH buffers 4 and 7, deionized water, instructions, 9V battery, hard carrying case. Kit Dimensions: 11" x 9.5" x 2"
Catalog No. 50-192-9321 Supplier Electron Microscopy Sciences Supplier No. 7208702
Includes everything necessary for immediate field use: Basic pH Meter, epoxy body, sealed reference combination electrode with a BNC connector and 2.5-ft cable, pH buffers 4 and 7, deionized water, instructions, 9V battery, hard carrying case. Kit Dimensions: 11" x 9.5" x 2"