Learn More
Cole-Parmer Oakton pH 5+ Handheld Meter Kit with Case, Solutions, and pH/ATC Probe
Catalog No. 501961346 Shop All Cole-Parmer Products
Encompass_Preferred
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.
Catalog No. 50-196-1346 Supplier Cole-Parmer Supplier No. 3561354
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.