Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ pH Meter SevenDirect SD20

Catalog No. 01911920 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
SevenDirect SD20 HA Kit
SevenDirect SD20 Kit
SevenDirect SD20 LowVol Kit
SevenDirect SD20 Organic Kit
SevenDirect SD20 Pure H2O Kit
SevenDirect SD20 Solids Kit
SevenDirect SD20 Stirrer Kit
SevenDirect SD20 Viscous Kit
SevenDirect SD20, Meter Only
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-911-920 SevenDirect SD20 Stirrer Kit
01-911-909 SevenDirect SD20, Meter Only
01-911-913 SevenDirect SD20 Kit
01-911-914 SevenDirect SD20 HA Kit
01-911-915 SevenDirect SD20 Organic Kit
01-911-916 SevenDirect SD20 Pure H2O Kit
01-911-917 SevenDirect SD20 Solids Kit
01-911-918 SevenDirect SD20 Viscous Kit
01-911-919 SevenDirect SD20 LowVol Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 01-911-920 Supplier Mettler Toledo™ Supplier No. 30671555
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The SevenDirect SD20 is an accurate and flexible benchtop meter for the determination of pH and ORP. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
TRUSTED_SUSTAINABILITY

Specifications

Includes Instrument, power s upply, protective cover, Get Started guide, electrode holder EasyPlace, appropriate electrode, calibration solutions, EasyMix magnatic stirrer
Accuracy (Temperature) ±0.1°C between 0°C and 100°C
Accuracy (mV) 0.1 mV
Accuracy (pH) 0.002
Data Management Details PC, Printer, USB Stick
Measurement Modes pH with mV and Temperature
Calibration Linear/Segmented
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) -2000 to 2000 mV
Resolution (mV) 1, 0.1 mV
Calibration Points Up to 5
Temperature Range (Metric) -30°C to +130°C
Temperature Range (English) -22°F to +266°F
Resolution (Temperature) 0.1°C
Display Type Color TFT
Backlight Option Yes
Memory 2000 Standard Measurements
Data Points 2000
Log Function Type Automatic, Manual
IP Rating IP54
Dimensions (L x W x H) 195 x 205 x 65 mm
Weight (Metric) 850 g
Weight (English) 1.9 lb.
Channels 1
Temperature Sensor InLab Expert-Pro ISM pH/Temperature Sensor
Type SevenDirect SD20 Stirrer Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.