Learn More
Chemglass Life Sciences pH Meter, Stainless Steel Enclosure, Digital Display, 115V, with Output to Record Data, Supplied with CG-1872-M-25
SDP
Catalog No. 501539694
Encompass_Preferred
Digital display meters/monitors for use with CG-1872 pH electrodes with an integrated PT100 sensor for temperature correction. Connection is made using a standard BNC probe connector with a 3-pin banana connector plug for RTD. Supplied with CG-1872-M-25 Adapter Cables. Please Note: Probes are NOT Included with Meters.
Catalog No. 50-153-9694 Supplier Chemglass Life Sciences Supplier No. CG1872M40
Digital display meters/monitors for use with CG-1872 pH electrodes with an integrated PT100 sensor for temperature correction. Connection is made using a standard BNC probe connector with a 3-pin banana connector plug for RTD. Supplied with CG-1872-M-25 Adapter Cables. Please Note: Probes are NOT Included with Meters.