Learn More
Chemglass Life Sciences Chemglass pH Meter, Stainless Steel Enclosure, Digital Display, 115 V (CG-1872-M-20)
SDP
Catalog No. 502733573
Encompass_Preferred
The Chemglass digital pH meter is designed for process reactor use and is housed in a stainless steel enclosure. It provides a clear digital readout and supports temperature compensation via an integrated PT100 input. The meter connects to pH electrodes through a standard BNC probe connector and is supplied with an adapter cable for easy installation.
  • Stainless steel enclosure for corrosion resistance
  • Digital display with easy-to-read readout
  • Integrated PT100 input for temperature compensation
  • Standard BNC probe connector for electrode connection
  • Supplied with adapter cable for probe and RTD connections
  • 115 V power configuration
Catalog No. 50-273-3573 Supplier Chemglass Life Sciences Supplier No. CG1872M20
Add to Cart
Add to Cart
The Chemglass digital pH meter is designed for process reactor use and is housed in a stainless steel enclosure. It provides a clear digital readout and supports temperature compensation via an integrated PT100 input. The meter connects to pH electrodes through a standard BNC probe connector and is supplied with an adapter cable for easy installation.
  • Stainless steel enclosure for corrosion resistance
  • Digital display with easy-to-read readout
  • Integrated PT100 input for temperature compensation
  • Standard BNC probe connector for electrode connection
  • Supplied with adapter cable for probe and RTD connections
  • 115 V power configuration