Learn More
Myron L Company PH METER TECH PRO2 TPH1
SDP
Catalog No. NC9660203
Encompass
Meas Conductivity, TDS, pH & temp. Dig. handheld. Item is sold with a non-dated COA. If dated COA is needed please also order Fisher part # NC9672510, which is the $40 COA fee. If request is made for dated COA after order is placed there will be a $70 fee.
Catalog No. NC9660203 Supplier Myron L Company Supplier No. TPH1
Meas Conductivity, TDS, pH & temp. Dig. handheld. Item is sold with a non-dated COA. If dated COA is needed please also order Fisher part # NC9672510, which is the $40 COA fee. If request is made for dated COA after order is placed there will be a $70 fee.