Learn More
Cole-Parmer Oakton 220 pH/ORP/Temperature Controller, 1/8 DIN
Supplier:  Cole-Parmer 5670015
Encompass_Preferred
Oakton pH/ORP controller features automatic buffer recognition, absolute and relative ORP measurements, and automatic temperature compensation to ensure measurement accuracy. Advanced features include two sets of programmable on/off relays are ideal for high/low control to activate an alarm, a pump, or another device and an isolated 4-20 mA output signal are suitable for opening valves and pacing pumps. ATC element is user selectable, 10K NTC or 1000 Ohm RTD. Order 56105-60 BNC-to-Stripped Wire Adapter for easy connections on terminal strip on back of controller.
Catalog No. 50-196-1389