Learn More
Azzota Corp Ph/Orp Testrode - All-In-One Tester,Large, Easy-To-Read Backlit Display,Standard 12mm Replaceable Electrode,1, 2 Or 3-Point Calibration,+/-0.02 Ph Unit Accuracy
Catalog No. 501612912
Encompass_Preferred
pH/ORP TesTrode - All-in-One Tester,Large, Easy-to-Read Backlit Display,Standard 12mm Replaceable Electrode,1, 2 or 3-Point Calibration,+/-0.02 pH Unit Accuracy
Catalog No. 50-161-2912 Supplier Azzota Corp Supplier No. POTT
pH/ORP TesTrode - All-in-One Tester,Large, Easy-to-Read Backlit Display,Standard 12mm Replaceable Electrode,1, 2 or 3-Point Calibration,+/-0.02 pH Unit Accuracy