Learn More
Grainger pH/ORP Transmitter w/Display ABS
Catalog No. 502767959
Encompass_Preferred
The SENSOREX TX100 is a pH and ORP transmitter with a 4-20 mA output signal. It features a rugged, waterproof enclosure and is designed for continuous monitoring and control applications. The TX100 has a measurement range of 0-14 pH and ±2000 mV ORP. It provides temperature compensation from 0-100°C. This transmitter can be easily calibrated and configured in the field using the built-in buttons and digital display. The TX100 is ideal for monitoring pH and ORP in industrial processes, municipal water treatment, food and beverage, and other applications requiring a reliable 4-20 mA measurement signal.
Catalog No. 50-276-7959 Supplier Grainger Supplier No. 16Z037
The SENSOREX TX100 is a pH and ORP transmitter with a 4-20 mA output signal. It features a rugged, waterproof enclosure and is designed for continuous monitoring and control applications. The TX100 has a measurement range of 0-14 pH and ±2000 mV ORP. It provides temperature compensation from 0-100°C. This transmitter can be easily calibrated and configured in the field using the built-in buttons and digital display. The TX100 is ideal for monitoring pH and ORP in industrial processes, municipal water treatment, food and beverage, and other applications requiring a reliable 4-20 mA measurement signal.