Learn More
Hach Company pH paper-1.0 - 11.0 pH range
Supplier:  Hach Company 39133
Encompass
Provides simple fast economical method of pH measurement Sharp color change for each 0 5 pH unit Convenient for sample pH adjustments Recommended for sewage water not finished water Five 4 6-meter (15 ) rolls per package
Catalog No. 50-002-56970