Learn More
Hach Company pH Paper, 3.0 - 5.5 pH Range
Catalog No. 501994263 Shop All Hach Company Products
Encompass
Provides simple, fast, economical method of pH measurement. Sharp color change for each 0.5 pH unit. Convenient for sample pH adjustments. Recommended for sewage water, not finished water. Five 4.6-meter (15') rolls per package.
Catalog No. 50-199-4263 Supplier Hach Company Supplier No. 37333
Provides simple, fast, economical method of pH measurement. Sharp color change for each 0.5 pH unit. Convenient for sample pH adjustments. Recommended for sewage water, not finished water. Five 4.6-meter (15') rolls per package.