Learn More
Hach Company pH paper-9.0 - 12.0 pH range
Supplier: Hach Company 38533
Provides simple fast economical method of pH measurement Sharp color change for each 0 5 pH unit Convenient for sample pH adjustments Recommended for sewage water not finished water Five 4 6-meter (15 ) rolls per package