Learn More
Med Vet International pH Paper Indicator Strips, pH Range 5-9, J0893M, 100 Strips/Vial
Catalog No. 5000582203
Encompass_Preferred
pH Paper Individual Strips in a vial. pH range 5-9, colors for each 0.5 pH unit. 100 strips/vial.
Catalog No. 50-005-82203 Supplier Med Vet International Supplier No. JOR893M
pH Paper Individual Strips in a vial. pH range 5-9, colors for each 0.5 pH unit. 100 strips/vial.