Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ pH Paper Strips
SCI_ED

Catalog No. S22515 Shop All Fisherbrand Products
Change view
Click to view available options
pH Range:
0 to 14
0 to 3
10 to 12
3 to 5.5
6 to 8
8 to 9.5
Type:
pH Test Paper
pH Test Paper Set
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range Type
S22515 0 to 14 pH Test Paper Set
S22508 0 to 14 pH Test Paper
S22509 3 to 5.5 pH Test Paper
S22510 6 to 8 pH Test Paper
S22511 0 to 3 pH Test Paper
S22512 8 to 9.5 pH Test Paper
S22513 10 to 12 pH Test Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. S22515

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An easy choice for classroom experiments or demonstrations

  • Ideal for educational use
  • Low-cost paper strips are easy to pass around the classroom
  • Easy-to-usedash (em)simply dip strip in solution to be tested and match to color chart immediately

Order Info

Box of 10 vials, 100 low-cost paper strips per vial.

Specifications

pH Range 0 to 14
For Use With (Application) For classroom experiments or demonstrations
Includes 1 vial each of 13-640-511, 13-640-509, 13-640-512, 13-640-513, and 13-640-514. 2 vials of 13-640-510, 3 vials of 13-640-508
Quantity 100 Tests/Vial, 10 Vials/Cs.
Type pH Test Paper Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.