Learn More
Description
- Ideal for educational use
- Low-cost paper strips are easy to pass around the classroom
- Easy-to-usedash (em)simply dip strip in solution to be tested and match to color chart immediately
Order Info
Box of 10 vials, 100 low-cost paper strips per vial.
Specifications
Specifications
| pH Range | 8 to 9.5 |
| For Use With (Application) | For classroom experiments or demonstrations |
| Quantity | 100 Tests/Vial, 10 Vials/Cs. |
| Type | pH Test Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
