Learn More
Medchemexpress LLC Ph-PEG3 100mg
SDP
Catalog No. 5000205556
Encompass_Preferred
Ph-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05556 Supplier Medchemexpress LLC Supplier No. HYW096158100MG
Ph-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]