Learn More
MSC Zep Neutral Floor Cleaner, pH Perfect, Low-Foam, 55 gal
Catalog No. 502815962
Encompass
Zep pH Perfect Neutral Floor Cleaner is a low-foam, general-purpose cleaner with a neutral pH (7-8), designed for most flooring surfaces. It is excellent for automatic floor scrubbers, rinsing easily to leave a film-free surface that will not harm floor finishes.
  • Controlled alkalinity with powerful cleaning action for manual and automatic scrubbing
  • Cleans and leaves a pleasant, fresh fragrance behind
  • Versatile for cleaning equipment, walls, and work surfaces
  • Highly concentrated with dilutions of 0.5 oz to 2 oz per gallon of water
  • Contains biodegradable detergents and emulsifiers
Catalog No. 50-281-5962 Supplier MSC Supplier No. 12339644
Zep pH Perfect Neutral Floor Cleaner is a low-foam, general-purpose cleaner with a neutral pH (7-8), designed for most flooring surfaces. It is excellent for automatic floor scrubbers, rinsing easily to leave a film-free surface that will not harm floor finishes.
  • Controlled alkalinity with powerful cleaning action for manual and automatic scrubbing
  • Cleans and leaves a pleasant, fresh fragrance behind
  • Versatile for cleaning equipment, walls, and work surfaces
  • Highly concentrated with dilutions of 0.5 oz to 2 oz per gallon of water
  • Contains biodegradable detergents and emulsifiers