Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System

Catalog No. 1329867 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Type:
pH Probe
pH and pH/ORP probe
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-298-67 pH Probe
13-298-66 pH and pH/ORP probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-298-67 Supplier YSI™ Supplier No. 005564
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Measure pH and pH & ORP/Redox quickly and easily

  • User-replaceble sensor allows for quick sensor replacements if needed
  • No tools necessary

TRUSTED_SUSTAINABILITY

Specifications

Type pH Probe
For Use With (Application) Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications
For Use With (Equipment) YSI model 556 handheld multi probe meter
Includes Storage bottle with storage solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.