Learn More
Chemglass Life Sciences 12mm Shaft Diameter, 125mm Usable Shaft Length, 25mm Minimum Immersion Length, Glass Shaft Material, Plug In Type G Electrode Head, 0 to 100°C Temperature Range
SDP
Catalog No. CG187203
Encompass_Preferred
Metrohm pH electrodes designed to be used with Chemglass reactors and data loggers. All electrodes are supplied with integrated PT 1000 sensor for temperature correction and have a BNC connector. 12mm Shaft Diameter, 125mm Usable Shaft Length, 25mm Minimum Immersion Length, Glass Shaft Material, Plug In Type G Electrode Head, 0 to 100¿C Temperature Range.Electrode is designed to fit into a CG-1872-B baffle using the CG-1872-A-10 adapter.
Catalog No. CG187203 Supplier Chemglass Life Sciences Supplier No. CG187203
Metrohm pH electrodes designed to be used with Chemglass reactors and data loggers. All electrodes are supplied with integrated PT 1000 sensor for temperature correction and have a BNC connector. 12mm Shaft Diameter, 125mm Usable Shaft Length, 25mm Minimum Immersion Length, Glass Shaft Material, Plug In Type G Electrode Head, 0 to 100¿C Temperature Range.Electrode is designed to fit into a CG-1872-B baffle using the CG-1872-A-10 adapter.