Learn More
Chemglass Life Sciences pH Probe, with K8, 12 x 425mm
SDP
Catalog No. 50143161
Encompass
pH Range: 0-12 Electrodes are filled with gel reference electrolyte under a pressure of 2. 5 bar Pressure Range: From vacuum up to a process of maximum 36PSI (2. 5 bar) Temperature Range: Steam sterilization/autoclavable to 266 degreesF (0 to 130 degreesC) In-situ sterilization: Up to 100 cycles under standard conditions Fully autoclavable or in-situ sterializable
Catalog No. 50-143-161 Supplier Chemglass Life Sciences Supplier No. CLS143579
pH Range: 0-12 Electrodes are filled with gel reference electrolyte under a pressure of 2. 5 bar Pressure Range: From vacuum up to a process of maximum 36PSI (2. 5 bar) Temperature Range: Steam sterilization/autoclavable to 266 degreesF (0 to 130 degreesC) In-situ sterilization: Up to 100 cycles under standard conditions Fully autoclavable or in-situ sterializable