Learn More
Bellco Glass, Inc. pH Probe,12x200mm Mettler
SDP
Catalog No. 501946284
Encompass_Preferred
Electrodes are filled with gel reference electrolyte under a pressure of 25 BAR pressure range from vacuum up to a process of max 36 PSI 25 BAR. Temperature range steam sterilization autoclave to 266F to 130C. In-Situ sterilization up to 100 cycles under standard conditions. Gel electrolyte eliminates fill requirements to reduce maintenance. Patented silver-ion trap prevents sulfide-bearing poisoning of the liquid junction increasing electrode life even in sulfide-bearing media suitable for direct insertion through compression fittings or top or side mounting in a stationary or retractable housing. Engraved serial number on each electrode for easy identification when working under GMP.
Catalog No. 50-194-6284 Supplier Bellco Glass, Inc. Supplier No. 780321510
Electrodes are filled with gel reference electrolyte under a pressure of 25 BAR pressure range from vacuum up to a process of max 36 PSI 25 BAR. Temperature range steam sterilization autoclave to 266F to 130C. In-Situ sterilization up to 100 cycles under standard conditions. Gel electrolyte eliminates fill requirements to reduce maintenance. Patented silver-ion trap prevents sulfide-bearing poisoning of the liquid junction increasing electrode life even in sulfide-bearing media suitable for direct insertion through compression fittings or top or side mounting in a stationary or retractable housing. Engraved serial number on each electrode for easy identification when working under GMP.