Learn More
Broadley James Corporation F-615 Sterilizable pH Electrode 130mm Length
Catalog No. NC9398012
Encompass
The F-615 style offers a T-pull handle which makes installation and cable routing easy in large tanks. Typically used in steel vessels 20 liters and up, this design is very robust and withstands multiple SIP and CIP cycles. Steam sterilizable.
Catalog No. NC9398012 Supplier Broadley James Corporation Supplier No. F615B130DH
The F-615 style offers a T-pull handle which makes installation and cable routing easy in large tanks. Typically used in steel vessels 20 liters and up, this design is very robust and withstands multiple SIP and CIP cycles. Steam sterilizable.