Learn More
Chemglass Life Sciences pH Probe Cable K8 connection to BNC, 10'
SDP
Catalog No. 50143140
Encompass
pH Probe Cable K8 connection to BNC, 10'. Standard BNC connector to AK9 provides interface connection between pH probe and controller.
Catalog No. 50-143-140 Supplier Chemglass Life Sciences Supplier No. CLS1435105
pH Probe Cable K8 connection to BNC, 10'. Standard BNC connector to AK9 provides interface connection between pH probe and controller.