Learn More
Chemglass Life Sciences pH Probe Cable K8 connection to BNC, 3'
SDP
Catalog No. 50143138
Encompass
pH Probe Cable K8 connection to BNC, 3'. Standard BNC connector to AK9 provides interface connection between pH probe and controller. For use with CLS-1438-002.
Catalog No. 50-143-138 Supplier Chemglass Life Sciences Supplier No. CLS1435100
pH Probe Cable K8 connection to BNC, 3'. Standard BNC connector to AK9 provides interface connection between pH probe and controller. For use with CLS-1438-002.