Learn More
Chemglass Life Sciences pH Probe Cable K8 connection to BNC, 6'
SDP
Catalog No. 50143139
Encompass
pH Probe Cable K8 connection to BNC, 6'. Standard BNC connector to AK9 provides interface connection between pH probe and controller. For use with CLS-1438-002.
Catalog No. 50-143-139 Supplier Chemglass Life Sciences Supplier No. CLS1435103
pH Probe Cable K8 connection to BNC, 6'. Standard BNC connector to AK9 provides interface connection between pH probe and controller. For use with CLS-1438-002.