Learn More
Bellco Glass, Inc. pH Probe Tester, 9V
SDP
Catalog No. 501946292
Encompass_Preferred
Handheld unit designed for testing pH, redox and ORP probes prior transmitter calibration. LCD display shows the absolute mV value to qualify probe status. Zero and span ranges are verified with 4.00 7.00 and 10.00 buffers. Probes can be tested in-situ during operation while maintaining sterility. Battery test insures proper operation. Replaceable 9V battery. Power requirement 9v battery suppliedRange 0-1000 mvResolution 1mvAccuracy 1mvInput impedance 6x10 to the 12th omhsDisplay type LCDBattery test 100 9vBattery range 80-110Battery nominal draw 20ma maximumReplace battery if test reading falls below
Catalog No. 50-194-6292 Supplier Bellco Glass, Inc. Supplier No. 780321750
Handheld unit designed for testing pH, redox and ORP probes prior transmitter calibration. LCD display shows the absolute mV value to qualify probe status. Zero and span ranges are verified with 4.00 7.00 and 10.00 buffers. Probes can be tested in-situ during operation while maintaining sterility. Battery test insures proper operation. Replaceable 9V battery. Power requirement 9v battery suppliedRange 0-1000 mvResolution 1mvAccuracy 1mvInput impedance 6x10 to the 12th omhsDisplay type LCDBattery test 100 9vBattery range 80-110Battery nominal draw 20ma maximumReplace battery if test reading falls below