Learn More
Bellco Glass, Inc. pH Probe W/VP,12x325mm With VP connector - Ingold
SDP
Catalog No. 501946296
Encompass_Preferred
Mettler Toledo ph probe – inpro seriesLow-maintenance ph-electrodesWith vario pin connectionMounting diameter 12mmInsertion length 325mmPh range 012Temperature range 140cPressure resistance bar up to 4 bar over pressurePressure resistance psi up to 58psi over-pressure--Diaphragm ceramic
Catalog No. 50-194-6296 Supplier Bellco Glass, Inc. Supplier No. 780322325
Mettler Toledo ph probe – inpro seriesLow-maintenance ph-electrodesWith vario pin connectionMounting diameter 12mmInsertion length 325mmPh range 012Temperature range 140cPressure resistance bar up to 4 bar over pressurePressure resistance psi up to 58psi over-pressure--Diaphragm ceramic