Learn More
Chemglass Life Sciences pH Probe, with VP, 12 x 225mm
SDP
Catalog No. 50143150
Encompass
pH probe with VP, 12x225mm. • pH Range: 0-12 • Electrodes are filled with gel reference electrolyte under a pressure of 2.5 bar • Pressure Range: From vacuum up to a process of maximum 36PSI (2.5 bar) • Temperature Range: Steam sterilization/autoclavable to 266°F (0 to 130°C) • In-situ sterilization: Up to 100 cycles under standard conditions • Fully autoclavable or in-situ sterializable
Catalog No. 50-143-150 Supplier Chemglass Life Sciences Supplier No. CLS143533
pH probe with VP, 12x225mm. • pH Range: 0-12 • Electrodes are filled with gel reference electrolyte under a pressure of 2.5 bar • Pressure Range: From vacuum up to a process of maximum 36PSI (2.5 bar) • Temperature Range: Steam sterilization/autoclavable to 266°F (0 to 130°C) • In-situ sterilization: Up to 100 cycles under standard conditions • Fully autoclavable or in-situ sterializable