Learn More
Chemglass Life Sciences pH Probe, with VP, 12 x 325mm
SDP
Catalog No. 50143151
Encompass
pH probe with VP, 12x325mm. • pH Range: 0-12 • Electrodes are filled with gel reference electrolyte under a pressure of 2.5 bar • Pressure Range: From vacuum up to a process of maximum 36PSI (2.5 bar) • Temperature Range: Steam sterilization/autoclavable to 266°F (0 to 130°C) • In-situ sterilization: Up to 100 cycles under standard conditions • Fully autoclavable or in-situ sterializable
Catalog No. 50-143-151 Supplier Chemglass Life Sciences Supplier No. CLS143535
pH probe with VP, 12x325mm. • pH Range: 0-12 • Electrodes are filled with gel reference electrolyte under a pressure of 2.5 bar • Pressure Range: From vacuum up to a process of maximum 36PSI (2.5 bar) • Temperature Range: Steam sterilization/autoclavable to 266°F (0 to 130°C) • In-situ sterilization: Up to 100 cycles under standard conditions • Fully autoclavable or in-situ sterializable