Learn More
Eurofins Discoverx PathHunter ProLabel Detection Kit, 10,000 dp (384-well). (93-0180L)
Catalog No. 501999878
Encompass
The PathHunter ProLabel Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cells overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 25 plates (384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0180l.
Catalog No. 50-199-9878 Supplier Eurofins Discoverx Supplier No. 930180L
Add to Cart
Add to Cart
Request a Quote
The PathHunter ProLabel Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cells overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 25 plates (384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0180l.