Learn More
Eurofins Discoverx PH PROLABEL DETECTN KIT 800DP
Catalog No. NC2394619
Encompass
The PathHunter® ProLabel Detection Kit is for use with PathHunter Cell Lines expressing ProLabel® (PL) or ProLink™ (PK) expression or cloning vectors. The kit measures the total amount of ProLabel- or ProLink-tagged protein expressed in cells. Functional assays using PathHunter Cell Lines expressing both a ProLabel- or ProLinktagged protein and EA require the PathHunter Detection Kit (93-0001). The assay can be used in both 96-well and 384-well microplate formats. 800 dp (384-well)
Catalog No. NC2394619 Supplier Eurofins Discoverx Supplier No. 930180
The PathHunter® ProLabel Detection Kit is for use with PathHunter Cell Lines expressing ProLabel® (PL) or ProLink™ (PK) expression or cloning vectors. The kit measures the total amount of ProLabel- or ProLink-tagged protein expressed in cells. Functional assays using PathHunter Cell Lines expressing both a ProLabel- or ProLinktagged protein and EA require the PathHunter Detection Kit (93-0001). The assay can be used in both 96-well and 384-well microplate formats. 800 dp (384-well)