Learn More
Inorganic Ventures pH 4 RED CALIBRATION STD 500m
SDP
Catalog No. NC1018010
Encompass_Preferred
Inorganic Ventures' pH 4 Red Standard is a 500mL certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration .
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1018010 Supplier Inorganic Ventures Supplier No. PHRED4500ML
Inorganic Ventures' pH 4 Red Standard is a 500mL certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration .
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search