Learn More
Hach Company pH Sen-PPS-5-Preamp-Convert
Catalog No. 5000207813 Shop All Hach Company Products
Encompass
pH Sensor-PPS Body-5 Wire with Built-in Preamplifier-Convertible Style-20-ft CablePPS Convertible Mounting Style 6m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode 20 ft Cable
Catalog No. 50-002-07813 Supplier Hach Company Supplier No. 6022P020
pH Sensor-PPS Body-5 Wire with Built-in Preamplifier-Convertible Style-20-ft CablePPS Convertible Mounting Style 6m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode 20 ft Cable