Learn More
Hach Company pH Sens-LCP-5 Wire-Convert-Fsh
Catalog No. 5000208093 Shop All Hach Company Products
Encompass
pH Sensor-LCP Body-5 Wire-Convertible-Flush ElectrodeLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Flush pH Glass Electrode
Catalog No. 50-002-08093 Supplier Hach Company Supplier No. 6028P510
pH Sensor-LCP Body-5 Wire-Convertible-Flush ElectrodeLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Flush pH Glass Electrode