Learn More
Hach Company pH Sens-LCP-5 Wire-Flow-Thru
Catalog No. 5000207859 Shop All Hach Company Products
Encompass
pH Sensor-LCP Body-5 Wire-Flow-Thru Mount-25-ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 7 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07859 Supplier Hach Company Supplier No. 6058P025
pH Sensor-LCP Body-5 Wire-Flow-Thru Mount-25-ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 7 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode