Learn More
Hach Company pH Sens-PPS-5 Wire-Flow-Thru
Catalog No. 5000207852 Shop All Hach Company Products
Encompass
pH Sensor-PPS Body-5 Wire-Flow-Thru Mount-10-ft CablePPS Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07852 Supplier Hach Company Supplier No. 6052P0
pH Sensor-PPS Body-5 Wire-Flow-Thru Mount-10-ft CablePPS Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode