Learn More
Hach Company pH Sens-Union-LCP-2-wire 4-20
Catalog No. 5000207436 Shop All Hach Company Products
Encompass
pH Sensor-Union-mount-LCP (Liquid Crystal Polymer) Body Material-2-wire with 4-20mA Output-Glass ElectrodeLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter General Purpose Glass pH Electrode
Catalog No. 50-002-07436 Supplier Hach Company Supplier No. 6458P0
pH Sensor-Union-mount-LCP (Liquid Crystal Polymer) Body Material-2-wire with 4-20mA Output-Glass ElectrodeLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter General Purpose Glass pH Electrode