Learn More
Hach Company pH Sens. UnionMt LCP w/Preamp
Catalog No. 5000207853 Shop All Hach Company Products
Encompass
pH Sensor-Union Mount-LCP (Liquid Crystal Polymer)-5-wire with Internal Preamplifier-10 ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07853 Supplier Hach Company Supplier No. 6058P0
pH Sensor-Union Mount-LCP (Liquid Crystal Polymer)-5-wire with Internal Preamplifier-10 ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode