Learn More
Sensorex pH Sensor, ATC, 12mm
Catalog No. 501897528
Encompass_Preferred
Sensorex 3-in-1 Comb pH/ATC Elect, 30K ATC element, BNC/8PIN DIN, can be used with Orion Series A/Hach EC
Catalog No. 50-189-7528 Supplier Sensorex Supplier No. S290C
Sensorex 3-in-1 Comb pH/ATC Elect, 30K ATC element, BNC/8PIN DIN, can be used with Orion Series A/Hach EC