Learn More
Sensorex pH Sensor,ATC,12mm,Mettler,P1K
Catalog No. 501897536
Encompass_Preferred
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and automatic temperature compensation 1000 Ohm (ATC) w/ DIN M + 3.5mm phono plug, use with Mettler Toledo.
Catalog No. 50-189-7536 Supplier Sensorex Supplier No. S299C
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and automatic temperature compensation 1000 Ohm (ATC) w/ DIN M + 3.5mm phono plug, use with Mettler Toledo.