Learn More
Sensorex pH Sensor,ATC,12mm,Orion EA/SA
Catalog No. 501897529
Encompass_Preferred
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and automatic temperature compensation (ATC). Can be used with Orion EA/SA, 30K ATC, BNC+Banana Plug x 2
Catalog No. 50-189-7529 Supplier Sensorex Supplier No. S291C
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and automatic temperature compensation (ATC). Can be used with Orion EA/SA, 30K ATC, BNC+Banana Plug x 2