Learn More
Sensorex pH Sensor,ATC,12mm,Polymer
Catalog No. 501897533
Encompass_Preferred
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and 30K automatic temperature compensation (ATC) w/BNC+C3 connectors.
Catalog No. 50-189-7533 Supplier Sensorex Supplier No. S294C
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and 30K automatic temperature compensation (ATC) w/BNC+C3 connectors.