Learn More
Sensorex pH Sensor, ATC,BNC/2.5mm Phono
Catalog No. 501897532
Encompass_Preferred
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and automatic temperature compensation (ATC).
Catalog No. 50-189-7532 Supplier Sensorex Supplier No. S293C/30K/BNC/2.5
Sensorex 3-in-1 combination electrodes are excellent choices for applications requiring pH, reference, and automatic temperature compensation (ATC).