Learn More
Sensorex pH Sensor,Glass,DJ,Refill, BNC
Catalog No. 501897523
Encompass_Preferred
Our pH2400 is refillable and has a double junction reference to prevent fouling by contaminants that interact with Silver (Ag).
Catalog No. 50-189-7523 Supplier Sensorex Supplier No. SG200CD
Our pH2400 is refillable and has a double junction reference to prevent fouling by contaminants that interact with Silver (Ag).