Learn More
Description
- For the toughest conditions: PEEK shaft with excellent chemical and mechanical resistance and a real heavy-duty sensor
- No junction, no clogging: Due to the solid XEROLYT™ reference electrolyte, no membrane junction is needed, which eliminates the risk of clogging
- Always the right temperature: The integrated temperature sensor guarantees measurements are always performed with the correct temperature compensation
- DES™ - Diagnosable Efficient Sustainable: Increased efficiency with sensor diagnostics: faster problem solving, less downtime, maximized sensor lifespan, and support of preventive maintenance
- Stay ahead: The sensor's diagnostic capability means peace of mind for the sensor condition. It supports preventive maintenance, maximizes the sensor's lifespan, and prevents premature discard
- Sustainable investment: DES™ digital sensors are a smart choice. Retaining the measurement electronics and only replacing the sensor shaft at the end of its life, or the detachable cable that can be used for all sensors, helps reduce material waste and thus, the ecological footprint
- Boost productivity and efficiency: With diagnosed errors, the user guidance with troubleshooting recommendations allows for faster problem-solving, thus reducing downtime. It also minimizes handling errors, therefore cutting down on the need for specialized training
- For use with METTLER TOLEDO NineFocus. Connection cables and signal bridges may be required.
- Accessories and Care
- Open Junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Specifications
| Connector Types | Direct |
| Junction | Open |
| Material | PEEK |
| pH Range | 0 to 14 |
| Type | Combination pH Electrode |
| Internal Reference | ARGENTHAL™ with Ag+-trap |
| Membrane | U |
| Probe | NTC 30 kΩ |
| Reference Electrolyte | XEROLYT™ Polymer |
| Sample Compatibility | Beer, Suspended Soil, Water |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.